missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM213B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569890
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 86%.
FAM213B is a protein coding gene.
Specifications
| FAM213B | |
| Polyclonal | |
| Unconjugated | |
| FAM213B | |
| 2810405K02Rik; AI836168; C16H1orf93; C1orf93; C5H1orf93; Fam213b; family with sequence similarity 213 member B; family with sequence similarity 213, member B; hypothetical protein MGC26818; LOW QUALITY PROTEIN: prostamide/prostaglandin F synthase; peroxiredoxin like 2B; Peroxiredoxin-like 2B; PM/PGFS; Prostamide/PG F synthase; prostamide/PGF synthase; prostamide/prostaglandin F synthase; Protein FAM213B; prxl2; Prxl2b; RGD1308251; sb:cb782; selu3; wu:fc33a02; zgc:85644 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 127281 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TBF2 | |
| FAM213B | |
| synthetic peptide directed towards the middle region of human FAM213B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction