missing translation for 'onlineSavingsMsg'
Learn More

FAM212A Antibody, DyLight 405, Novus Biologicals™

Product Code. 18418673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418673 Supplier Novus Biologicals Supplier No. NBP176251V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM212A Polyclonal antibody specifically detects FAM212A in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM212A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias C3orf54, chromosome 3 open reading frame 54, family with sequence similarity 212, member A, INKA1, INKA1 protein, MGC20416
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the carboxy terminus of human FAM212A. The immunogen is located within the last 50 amino acids of FAM212A. Amino Acid Squence: CPPRPEPELPADVSR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 389119
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.