missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM20A Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | FAM20A |
|---|---|
| Dilution | Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228108
|
Novus Biologicals
NBP3-33384-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231905
|
Novus Biologicals
NBP3-33384-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM20A Monoclonal antibody specifically detects FAM20A in Human samples. It is validated for ELISA,Western BlotSpecifications
| FAM20A | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Stem Cells | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 54757 | |
| IgG | |
| Affinity purified |
| Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AIGFS, DKFZp434F2322, family with sequence similarity 20, member A, FP2747, UNQ9388/PRO34279 | |
| A synthetic peptide corresponding to a sequence within amino acids 500-600 of human FAM20A.2).,, Sequence:, AIFDFLIGNMDRHHYEMFTKFGDDGFLIHLDNARGFGRHSHDEISILSPLSQCCMIKKKTLLHLQLLAQADYRLSDVMRESLLEDQLSPVLTEPHLLALDR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title