missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM204A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546216
This item is not returnable.
View return policy
Description
Peptide sequence: NSGLNLQEDK EDESIRKTEI IDFSTDEPKT ETESNVNAYE ECPSGIPIDM Sequence homology: Dog: 79%; Horse: 79%; Human: 100%.
FAM204A is a protein coding gene.
Specifications
| FAM204A | |
| Polyclonal | |
| Unconjugated | |
| FAM204A | |
| bA319I23.1; C10orf84; FAM204A; family with sequence similarity 204 member A; family with sequence similarity 204, member A; protein FAM204A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 63877 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H8W3 | |
| FAM204A | |
| Synthetic peptide directed towards the N-terminal of human FAM204A (aa 27-76). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction