missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM204A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566447
This item is not returnable.
View return policy
Description
Immunogen sequence: QWKELTQYFGV NDRFDPPVKR KKVEKSGLEK RIDQAVEEWN IEKAEELSNQ L Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
FAM204A is a protein coding gene.
Specifications
| FAM204A | |
| Polyclonal | |
| Unconjugated | |
| FAM204A | |
| bA319I23.1; C10orf84; FAM204A; family with sequence similarity 204 member A; family with sequence similarity 204, member A; protein FAM204A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H8W3 | |
| FAM204A | |
| Recombinant Human FAM204A. Recombinant protein control fragment (Product #RP-107122). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction