missing translation for 'onlineSavingsMsg'
Learn More

FAM198A Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18417309 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18417309 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417309 Supplier Novus Biologicals Supplier No. NBP306029MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM198A Polyclonal antibody specifically detects FAM198A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM198A
Applications Immunocytochemistry
Classification Polyclonal
Dilution Immunocytochemistry/ Immunofluorescence
Gene Alias C3orf41, family with sequence similarity 198, member A, FLJ33682
Gene ID (Entrez) 729085
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.