missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM198A Polyclonal antibody specifically detects FAM198A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human FAM198A fragment. |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 729085 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?