missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM193A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560246
This item is not returnable.
View return policy
Description
Immunogen sequence: RSISTFLGTL ENEHLKKFQV TWELHNKHLF ENLVFSEPLL QSNLPALVSQ IRLGTTTHDT CSEDTYSTLL QRYQRSEEEL RRVAEEWLEC QKRIDAYVDE QM Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
FAM193A is a protein coding gene.
Specifications
| FAM193A | |
| Polyclonal | |
| Unconjugated | |
| FAM193A | |
| C4orf8; Fam193a; family with sequence similarity 193 member A; family with sequence similarity 193, member A; im:7155132; Protein FAM193A; Protein IT14; RES4-22; RGD1309634 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8603 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78312 | |
| FAM193A | |
| Recombinant protein corresponding to Human FAM193A. Recombinant protein control fragment (Product #RP-100143). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction