missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM189B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552318
This item is not returnable.
View return policy
Description
Immunogen sequence: LLLPRSHSDP GITTSSDTAD FRDLYTKVLE EEAASVSSAD TGLCSEACLF RLARCPSPKL LRARSAEKRR PVPTFQKVPL PSGPAPAHSL GDLKGSWPGR GLVTRFLQIS RKAPDPSGT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
This gene is located near the gene for the lysosomal enzyme glucosylceramidase; a deficiency in this enzyme is associated with Gaucher disease. The encoded protein has been identified as a potential binding partner of a WW domain-containing protein which is involved in apoptosis and tumor suppression. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2010].
Specifications
| FAM189B | |
| Polyclonal | |
| Unconjugated | |
| FAM189B | |
| 1110013L07Rik; C1orf2; COTE1; FAM189B; family with sequence similarity 189 member B; family with sequence similarity 189, member B; Protein COTE1; Protein FAM189B; RGD1306107 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10712, 310640, 68521 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P81408, Q5HZJ5 | |
| FAM189B | |
| Recombinant protein corresponding to Human FAM189B. Recombinant protein control fragment (Product #RP-89182). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction