missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM189B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58196-25ul
This item is not returnable.
View return policy
Description
FAM189B Polyclonal specifically detects FAM189B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| FAM189B | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| C1orf2, chromosome 1 open reading frame 2, COTE1, family with sequence similarity 189, member B, hypothetical protein LOC10712, Protein COTE1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10712 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| FAM189B | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PLYPTDCPPSYEAVMGLRGDSQATLFDPQLHDGSCICERVASIVDVSMDSGSLVLSAIGDLPGGSSPSEDSCLLELQGSVRSVDYVLFRSIQRSRAGYCLSLDCGLRGPFEESPLPRRPPRAARSYSCSA | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion