missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM184B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560616
This item is not returnable.
View return policy
Description
Immunogen sequence: ARQENSELKG TAKKLGEKLA VAKDRMMLQE CRGTQQTDAM KTELVSENKV LREENDLEAG NLHPQQDQSC LKECPCMKGG TDMQTKKEAS AETEYMKQQY EEDL Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 60%.
KIAA1276 was first identified as a hypothetical protein predicted from the in silico analysis of long cDNAs isolated in the Kazusa cDNA sequencing project. The function of KIAA1276 has not been characterized; however KIAA1276 homozygous knockout mice are viable.
Specifications
| FAM184B | |
| Polyclonal | |
| Unconjugated | |
| FAM184B | |
| FAM184B; family with sequence similarity 184 member B; family with sequence similarity 184, member B; KIAA1276; Protein FAM184B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULE4 | |
| FAM184B | |
| Recombinant protein corresponding to Human FAM184B. Recombinant protein control fragment (Product #RP-100189). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction