missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM184A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565124
This item is not returnable.
View return policy
Description
Immunogen sequence: YLMRESKPEDI QMITELKAML TERDQIIKKL IEDNKFYQLE LVNRETNFNK VFNSSPTVGV INPLAKQKKK NDKSPTNRFV SVPNLSALES GGVG Highest antigen sequence identity to the following orthologs - mouse 87%, rat 89%.
FAM184A (Family With Sequence Similarity 184 Member A) is a Protein Coding gene. An important paralog of this gene isFAM184B.
Specifications
| FAM184A | |
| Polyclonal | |
| Unconjugated | |
| FAM184A | |
| 3110012E06Rik; 4930438C08Rik; 4930589M24Rik; C6orf60; FAM184A; Fam184a-ps1; family with sequence similarity 184 member A; family with sequence similarity 184, member A; family with sequence similarity 184, member A, pseudogene 1; protein FAM184A; RGD1311294 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79632 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NB25 | |
| FAM184A | |
| Recombinant Human FAM184A. Recombinant protein control fragment (Product #RP-104645). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction