missing translation for 'onlineSavingsMsg'
Learn More

FAM181B Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18360367 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360367 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360367 Supplier Novus Biologicals Supplier No. NBP306306AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM181B Polyclonal specifically detects FAM181B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM181B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Family With Sequence Similarity 181, Member B, Protein FAM181B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM181B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220382
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.