missing translation for 'onlineSavingsMsg'
Learn More

FAM178B Polyclonal Antibody

Product Code. 15878663
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15878663 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15878663 Supplier Invitrogen Supplier No. PA570257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FAM178B Polyclonal Antibody

FAM178B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM178B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM178B
Gene Accession No. Q8IXR5
Gene Alias 1700024G10Rik; FAM178B; family with sequence similarity 178 member B; family with sequence similarity 178, member B; Gm990; HSPC234; Protein FAM178B; RGD1564591
Gene Symbols FAM178B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM178B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51252
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.