missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM176A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26726-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a regulator of programmed cell death, mediating both autophagy and apoptosis. Expressed in lung, kidney, liver, pancreas, placenta, but not in heart and skeletal muscle.Specifications
| FAM176A | |
| Polyclonal | |
| Unconjugated | |
| EVA1A | |
| FAM176A, Protein FAM176A, TMEM166, Transmembrane protein 166 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 232146, 84141 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91WM6, Q9H8M9 | |
| EVA1A | |
| FAM176A Fusion Protein Ag24990 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction