missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM175B Polyclonal Antibody

FAM175B Polyclonal Antibody
Brand: Invitrogen PA567326
This item is not returnable.
View return policy
Description
FAM175B Polyclonal Antibody for ICC/IF
Specifications
| FAM175B | |
| Polyclonal | |
| Unconjugated | |
| ABRAXAS2 | |
| AA589499; Abraxas brother; abraxas brother 1; abraxas brother protein 1; Abraxas2; Abro1; AI853413; BRISC complex subunit Abraxas 2; BRISC complex subunit Abro1; C430003P19Rik; Em:AC068896.4; FAM175B; family with sequence similarity 175 member B; family with sequence similarity 175, member B; KIAA0157; Protein FAM175B; RGD1308918 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15018 | |
| ABRAXAS2 | |
| Recombinant Human FAM175B. Recombinant protein control fragment (Product #RP-107971). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction