Learn More
Invitrogen™ FAM175B Monoclonal Antibody (K92017_12H1), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | FAM175B |
|---|---|
| Clone | K92017_12H1 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203849
|
Invitrogen™
MA555724 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30208693
|
Invitrogen™
MA556104 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: QEYKVSSVPN TSQSYAKVIK EHGTDFFDKD GVMKDIRAIY QVYNALQEKV QAVCADVEKS ERVVESCQAE VNKLRRQITQ RKNEKEQERR LQQAVLSRQM PSESLDPAFS PRMPSSGFAA EGRSTLGDAE ASDPPPPYSD FHPNNQESTL SHSRMERSVF MPRPQAVGSS NYASTSAGLK YPGSGADLPP PQRAAGDSGE DSDDSDYENL IDPTEPSNSE YSHSKDSRPM AHPDEDPRNT
FAM175B Monoclonal antibody specifically detects FAM175B in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| FAM175B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AA589499; Abraxas brother; abraxas brother 1; abraxas brother protein 1; Abraxas2; Abro1; AI853413; BRISC complex subunit Abraxas 2; BRISC complex subunit Abro1; C430003P19Rik; Em:AC068896.4; FAM175B; family with sequence similarity 175 member B; family with sequence similarity 175, member B; KIAA0157; Protein FAM175B; RGD1308918 | |
| Recombinant human ABRAXAS2 (immunogen range is 171-410). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ABRAXAS2 |
| K92017_12H1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q15018 | |
| 23172 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.