missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM173B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577209
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
FAM173B is a 233 amino acid single-pass membrane protein that is encoded by a gene that maps to human chromosome 5, which makes up about 6% of human genomic DNA and contains 181 million base pairs encoding around 1,000 genes. It is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
Specifications
| FAM173B | |
| Polyclonal | |
| Unconjugated | |
| FAM173B | |
| A930016P21Rik; AA987072; ATP synthase subunit C lysine N-methyltransferase; Atpsckmt; FAM173B; family with sequence similarity 173 member B; family with sequence similarity 173, member B; hFAM173B; JS-2; mFam173b; protein FAM173B; Protein N-lysine methyltransferase FAM173B; RGD1560629 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 134145, 499561, 68073 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P4H8, Q9D1Z3 | |
| FAM173B | |
| Recombinant full length Human FAM173B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction