missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM173B Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00134145-B01P
This item is not returnable.
View return policy
Description
FAM173B Polyclonal antibody specifically detects FAM173B in Human samples. It is validated for Western Blot
Specifications
| FAM173B | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_954584 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| family with sequence similarity 173, member B, FLJ20667, hypothetical protein LOC134145 | |
| LOC134145 (NP_954584, 1 a.a. - 233 a.a.) full-length human protein. MEGGGGIPLETLKEESQSRHVLPASFEVNSLQKSNWGFLLTGLVGGTLVAVYAVATPFVTPALRKVCLPFVPATMKQIENVVKMLRCRRGSLVDIGSGDGRIVIAAAKKGFTAVGYELNPWLVWYSRYRAWREGVHGSAKFYISDLWKVTFSQYSNVVIFGVPQMMLQLEKKLERELEDDARVIACRFPFPHWTPDHVTGEGIDTVWAYDASTFRGREKRPCTSMHFQLPIQA | |
| 0.05 mg | |
| Cell Biology | |
| 134145 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction