missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM171B Polyclonal antibody specifically detects FAM171B in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | FAM171B |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | family with sequence similarity 171, member B, FLJ34104, KIAA1946, NPD019 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human FAM171B (Accession#: AAH60872.1; Met1-Thr355) |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?