missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM170B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15710022
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15710022 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15710022 Supplier Invitrogen™ Supplier No. PA562222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PSSESSEYQS YSQYQSCCSC MCDEDNAAPQ SVCAFYTHVQ TVRGVAVAWE TEAGFEPVTR KPRIHEAQFI KRQRWNGS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

FAM170B plays a role in fertilization through the acrosome reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM170B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM170B
Gene Accession No. A6NMN3
Gene Alias 4922501K12Rik; Acrosome-related protein; AI449756; C10orf73; Fam170b; family with sequence similarity 170 member B; family with sequence similarity 170, member B; LOW QUALITY PROTEIN: protein FAM170B; protein FAM170B; putative protein FAM170B; rCG42349-like
Gene Symbols FAM170B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM170B. Recombinant protein control fragment (Product #RP-97158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170370
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.