missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM170A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558160
This item is not returnable.
View return policy
Description
Immunogen sequence: KRRQKRKHLE NEESQETAEK GGGMSKSQED ALQPGSTRVA KGWSQGVGEV TSTSEYCSCV SSSRKLIHSG IQRIHRDSPQ PQSPLAQVQE RGETPPRS Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.
Acts as a nuclear transcription factor that positively regulates the expression of heat shock genes. Binds to heat shock promoter elements (HSE).
Specifications
| FAM170A | |
| Polyclonal | |
| Unconjugated | |
| FAM170A | |
| FAM170A; family with sequence similarity 170 member A; family with sequence similarity 170, member A; protein FAM170A; Zinc finger domain-containing protein; zinc finger protein ZNFD; Znf domain containing protein; ZNFD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A1A519 | |
| FAM170A | |
| Recombinant protein corresponding to Human FAM170A. Recombinant protein control fragment (Product #RP-96961). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction