missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM168A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570083
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
In cancer context, protects cells from induced-DNA damage and apoptosis. Acts, at least in part, through PI3K/AKT/NFKB signaling pathway and by preventing POLB degradation. Decreases POLB ubiquitation and stabilizes its protein levels.
Specifications
| FAM168A | |
| Polyclonal | |
| Unconjugated | |
| FAM168A | |
| 2610030B18Rik; B930006L02Rik; FAM168A; family with sequence similarity 168 member A; family with sequence similarity 168, member A; KIAA0280; mKIAA0280; Protein FAM168A; RGD1308929; TCRP1; Tongue cancer chemotherapy resistance-associated protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 319604 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BGZ2 | |
| FAM168A | |
| synthetic peptide corresponding to a region of mouse FAM168A. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction