missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM160A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546454
This item is not returnable.
View return policy
Description
Peptide sequence: AVPSAPTGPG PLLEFALHED LLTRVLTWQL QWDELGDGVE ERRAEQLKLF.
FAM160A2 is a component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex).
Specifications
| FAM160A2 | |
| Polyclonal | |
| Unconjugated | |
| FAM160A2 | |
| C11orf56; FAM160A2; family with sequence similarity 160 member A2; family with sequence similarity 160, member A2; FHIP; FTS and Hook interacting protein; FTS and Hook-interacting protein; KIAA1759; Protein FAM160A2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84067 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N612 | |
| FAM160A2 | |
| Synthetic peptide directed towards the N-terminal of human FAM160A2 (aa 85-134). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction