missing translation for 'onlineSavingsMsg'
Learn More

FAM159A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15754461
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15754461 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15754461 Supplier Invitrogen Supplier No. PA556551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FAM159A Polyclonal Antibody

FAM159A Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM159A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Shisal2a
Gene Accession No. Q6UWV7
Gene Alias Fam159a; family with sequence similarity 159 member A; family with sequence similarity 159, member A; membrane protein FAM159A; OTTMUSG00000008243; PRO7171; Protein shisa-like-2A; Shisal2a; UNQ2783/PRO7171; WWLS2783
Gene Symbols Shisal2a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM159A. Recombinant protein control fragment (Product #RP-95717).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.