missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM156A Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97746PE
This item is not returnable.
View return policy
Description
FAM156A Polyclonal antibody specifically detects FAM156A in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| FAM156A | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| family with sequence similarity 156, member A, TMEM29 | |
| Produced in rabbits immunized with E. coli-derived Human FAM156A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 29057 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction