missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM156A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | FAM156A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM156A Polyclonal specifically detects FAM156A in Human samples. It is validated for Western Blot.Specifications
| FAM156A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| family with sequence similarity 156, member A, TMEM29 | |
| FAM156A | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_054857 | |
| 29057 | |
| Synthetic peptide directed towards the N terminal of human FAM156A. Peptide sequence DPLQKRNPASPSKSSPMTAAETSQEGPAPSQPSYSEQPMMGLSNLSPGPG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title