missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM155B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557517
This item is not returnable.
View return policy
Description
Immunogen sequence: SFSSNQGGGG LGLETLPALE EGLTREE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 93%.
TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ).
Specifications
| FAM155B | |
| Polyclonal | |
| Unconjugated | |
| FAM155B | |
| bB57D9.1; bB57D9.1 (TED protein); CXorf63; FAM155B; family with sequence similarity 155 member B; family with sequence similarity 155, member B; Protein TED; TED; TMEM28; Tmem28l; Transmembrane protein 28; transmembrane protein 28-like; transmembrane protein FAM155B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75949 | |
| FAM155B | |
| Recombinant protein corresponding to Human FAM155B. Recombinant protein control fragment (Product #RP-95443). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction