missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM154B Polyclonal specifically detects FAM154B in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | FAM154B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | DKFZp666G057, family with sequence similarity 154, member B, hypothetical protein LOC283726 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human FAM154B |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?