missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM133A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566826
This item is not returnable.
View return policy
Description
Immunogen sequence: KALAEFEEKMN ENWKKELEKS REKLLSG Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
FAM133A is a protein coding gene.
Specifications
| FAM133A | |
| Polyclonal | |
| Unconjugated | |
| FAM133A | |
| cancer/testis antigen 115; CT115; FAM133A; family with sequence similarity 133 member A; family with sequence similarity 133, member A; protein FAM133A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286499 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N9E0 | |
| FAM133A | |
| Recombinant Human FAM133A. Recombinant protein control fragment (Product #RP-105649). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction