missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM131A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560127
This item is not returnable.
View return policy
Description
Immunogen sequence: ELLLAKLPPS RESAFRSLGP LEAQDSLYNS PLTESCLSPA EEEPAPCKDC QPLCPPLTGS WERQRQASDL ASSGVVSL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.
Specifications
| FAM131A | |
| Polyclonal | |
| Unconjugated | |
| Fam131a | |
| 2900046G09Rik; C3orf40; Fam131a; family with sequence similarity 131 member A; family with sequence similarity 131, member A; FLAT715; PRO1378; protein FAM131A; UNQ715/PRO1378 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100360487, 131408, 78408 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXB0, Q8BWU3 | |
| Fam131a | |
| Recombinant protein corresponding to Human FAM131A. Recombinant protein control fragment (Product #RP-100235). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction