missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM129B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35917-20ul
This item is not returnable.
View return policy
Description
FAM129B Polyclonal antibody specifically detects FAM129B in Human samples. It is validated for ELISA,Western Blot
Specifications
| FAM129B | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| bA356B19.6, C9orf88, chromosome 9 open reading frame 88, DKFZP434H0820, family with sequence similarity 129, member B, FLJ13518, FLJ22151, FLJ22298, MEG-3, MINERVA, niban-like protein 1, OC58, Protein FAM129B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 447-746 of human FAM129B (NP_073744.2).,, Sequence:, TFETLLHQELGKGPTKEELCKSIQRVLERVLKKYDYDSSSVRKRFFREALLQISIPFLLKKLAPTCKSELPRFQELIFEDFARFILVENTYEEVVLQTVMKDILQAVKEAAVQRKHNLYRDSMVMHNSDPNLHLLAEGAPIDWGEEYSNSGGGGSPSPSTPESATLSEKRRRAKQVVSVVQDEEVGLPFEASPESPPPASPDGVTEIRGLLAQGLRPESPPPAGPLLNGAPAGESPQPKAAPEASSPPASPLQHLLPGKAVDLGPPKPSDQETGEQVSSPSSHPALHTTTEDSAGVQTEF | |
| 20 μL | |
| Cellular Signaling, Oncogenes | |
| 64855 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction