missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM127B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120572
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, A-549. Immunogen sequence: MDGRVQLMKA LLAGPLRPAA RRWRNPIPFP ETFDGDTDRL PEFIVQTSSY MFVDENTFSN DALKVTFLIT RLTGPALQWV IPYIRKESPL LNDYRGFLAE MKRVFGWEED EDF.
The function of this protein remains unknown.
Specifications
| FAM127B | |
| Polyclonal | |
| Unconjugated | |
| Rtl8a | |
| 1110012O05Rik; CAAX box 1 homolog A; CAAX box 1A; Cxx1a; CXX1B; FAM127B; family with sequence similarity 127 member B; family with sequence similarity 127, member B; LOC679038; mammalian retrotransposon derived 8b; mammalian retrotransposon derived protein 8A; Mar8.2A/B; MAR8A; Mart8b; protein FAM127B; Retrotransposon Gag-like protein 8A; RTL8A; similar to mammalian retrotransposon derived 8b; uncharacterized protein LOC679038 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26071, 66158 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.21 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BWD3 | |
| Rtl8a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human FAM127B (NP_001071640.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction