Learn More
Invitrogen™ FAM127A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120304
Description
Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, Lipid-anchor. Immunogen sequence: MDGRVQLIKA LLALPIRPAT RRWRNPIPFP ETFDGDTDRL PEFIVQTGSY MFVDENTFSS DALKVTFLIT RLTGPALQWV IPYIKKESPL LNDYRGFLAE MKRVFGWEED EDF.
RTL8C (Retrotransposon Gag Like 8C) is a Protein Coding gene. An important paralog of this gene is RTL8A.
Specifications
| FAM127A | |
| Polyclonal | |
| Unconjugated | |
| RTL8C | |
| 2900027G03Rik; CAAX box 1; CAAX box 1 homolog C; CAAX box 1C; CXX1; Cxx1c; FAM127A; family with sequence similarity 127 member A; family with sequence similarity 127, member A; mammalian retrotransposon derived 8a; mammalian retrotransposon derived protein 8C; MAR8; Mar8.1; MAR8C; MART8; Mart8a; MART8C; protein FAM127A; Retrotransposon Gag-like protein 8C; RTL8C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 72865 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.08 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| 0 | |
| RTL8C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human FAM127A (NP_001071639.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.