Learn More
Invitrogen™ FAM126A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534761
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (91%), Rat (89%), Rhesus Monkey (98%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.
Specifications
| FAM126A | |
| Polyclonal | |
| Unconjugated | |
| FAM126A | |
| AB030242; down regulated by Ctnnb1, a; down-regulated by CTNNB1 protein A; down-regulated by Ctnnb1, a; DRCTNNB1A; Fam126a; family with sequence similarity 126 member A; family with sequence similarity 126, member A; HCC; HLD5; HYCC1; Hyccin; Protein FAM126A; RGD1562906 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84652, 84668 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q6P9N1, Q9BYI3 | |
| FAM126A | |
| Recombinant protein encompassing a sequence within the center region of human FAM126A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.