missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM125A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25816-1-AP-20UL
This item is not returnable.
View return policy
Description
FAM125A, also named as MVB12A and CFBP, is a component of the ESCRT-I complex. FAM125A is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. FAM125A has three isoforms with MW 27-28 kDa and 25 kDa.Specifications
| FAM125A | |
| Polyclonal | |
| Unconjugated | |
| MVB12A | |
| CFBP, ESCRT I complex subunit MVB12A, FAM125A, Protein FAM125A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 93343 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96EY5 | |
| MVB12A | |
| FAM125A Fusion Protein Ag22919 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction