missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM125A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00
Specifications
| Antigen | FAM125A |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
FAM125A Polyclonal specifically detects FAM125A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FAM125A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 93343 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ISCTVEGAPASFGKSFAQKSGYFLCLSSLGSLENPQENVVADIQIVVDKSPLPLGFSPVCDPMDSKASVSKKKRMCVKLLPL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CFBP, CIN85/CD2AP family binding protein, CIN85/CD2AP family-binding protein, ESCRT-I complex subunit MVB12A, family with sequence similarity 125, member A, FLJ32495, multivesicular body subunit 12A, Protein FAM125A | |
| MVB12A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title