missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM120C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561568
This item is not returnable.
View return policy
Description
Immunogen sequence: KGHGKEQTGR GSKGHKKGNK QGSSDGVSKS LELHQGRSRS QVNGNSGALI KEEKSDHRLP APSQCALSRD SNECNNGNRY LPM Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
This gene encodes a potential transmembrane protein and lies in a region where mutations and deletions have been associated with intellectual disability and autism. Alternative splicing results in multiple transcript variants.
Specifications
| FAM120C | |
| Polyclonal | |
| Unconjugated | |
| FAM120C | |
| constitutive coactivator of PPAR-gamma-like protein 2; CXorf17; D930001I21Rik; FAM120C; family with sequence similarity 120, member C; family with sequence similarity 120C; ORF34; Protein FAM120C; RGD1564253; si:dkey-206f21.2; tumor antigen BJ-HCC-21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54954 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NX05 | |
| FAM120C | |
| Recombinant protein corresponding to Human FAM120C. Recombinant protein control fragment (Product #RP-100776). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction