missing translation for 'onlineSavingsMsg'
Learn More

FAM120B Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18401706 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401706 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401706 Supplier Novus Biologicals Supplier No. NBP176315AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM120B Polyclonal antibody specifically detects FAM120B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM120B
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CCPGKIAA0183, Constitutive coactivator of PPARG, Constitutive coactivator of PPAR-gamma, family with sequence similarity 120B, FLJ55614, KIAA1838constitutive coactivator of peroxisome proliferator-activated receptor gamma, PGCC1dJ894D12.1, PPARG constitutive coactivator 1, PPARgamma constitutive coactivator 1, Protein FAM120B
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human PGCC1. The immunogen is located within the last 50 amino acids of PGCC1. Amino Acid Squence: RDQGPGSRQYEHDQWRRY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84498
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.