missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM120A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA554069

Product Code. 15772001

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SGGATNHISG NKIGWEKTGS HSEPQARGDP GDQTKAEGSS TASSGSQLAE GKGSQMGTVQ PIPCLLSMPT RNHMDITTPP L Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

May participate in mRNA transport in the cytoplasm. Critical component of the oxidative stress-induced survival signaling. Activates src family kinases and acts as a scaffolding protein enabling src family kinases to phosphorylate and activate PI3-kinase. Binds RNA and promotes the secretion of IGF-II. May play a pivotal role in the progression of scirrhous-type gastric cancer by supporting cancer cell survival in environments with various oxidative stresses. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM120A
Polyclonal
Unconjugated
Fam120a
C9orf10; Constitutive coactivator of PPAR-gamma-like protein 1; DNAPTP1; DNAPTP5; Fam120a; family with sequence similarity 120, member A; family with sequence similarity 120A; Kiaa0183; Ossa; oxidative stess-associated Src activator; oxidative stress-associated Src activator; Protein FAM120A; RGD1306643
Rabbit
Antigen affinity chromatography
RUO
23196
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NZB2
Fam120a
Recombinant protein corresponding to Human FAM120A. Recombinant protein control fragment (Product #RP-92880).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.