missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM120A Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£444.00
Specifications
| Antigen | FAM120A |
|---|---|
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
FAM120A Polyclonal antibody specifically detects FAM120A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FAM120A | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C9orf10MGC133257, chromosome 9 open reading frame 10, constitutive coactivator of PPAR-gamma-like protein 1, DNA polymerase-transactivated protein 1, DNA polymerase-transactivated protein 5, DNAPTP1, DNAPTP5, family with sequence similarity 120A, KIAA0183MGC111527, OSSA, Oxidative stress-associated Src activator, Protein FAM120A | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the center of human FAM120A. The immunogen is located within amino acids 610 - 660 of FAM120A. Amino Acid Squence: SRKKTERLAFRKNRLPPE | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Cell Biology, Signal Transduction | |
| 50 mM Sodium Borate | |
| 23196 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title