missing translation for 'onlineSavingsMsg'
Learn More

FAM120A Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18409934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409934 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409934 Supplier Novus Biologicals Supplier No. NBP177367AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM120A Polyclonal antibody specifically detects FAM120A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM120A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C9orf10MGC133257, chromosome 9 open reading frame 10, constitutive coactivator of PPAR-gamma-like protein 1, DNA polymerase-transactivated protein 1, DNA polymerase-transactivated protein 5, DNAPTP1, DNAPTP5, family with sequence similarity 120A, KIAA0183MGC111527, OSSA, Oxidative stress-associated Src activator, Protein FAM120A
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the center of human FAM120A. The immunogen is located within amino acids 610 - 660 of FAM120A. Amino Acid Squence: SRKKTERLAFRKNRLPPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23196
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.