missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM117B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15779820
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15779820 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779820 Supplier Invitrogen™ Supplier No. PA551869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ECSPKQLHEI PAFYCPDKNK VNFIPKSGSA FCLVSILKPL LPTPDLTLKG SGHSLTVTTG MTTTLLQPIA VASLSTNTEQ DRVSRGTSTV MPSASLLPPP EPIEEAE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

FAM117B is encoded by ALS2CR13 which is mapped in the genomic region covering the complete candidate region for Amyotrophic lateral sclerosis 2 (ALS2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM117B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM117B
Gene Accession No. Q6P1L5
Gene Alias 2810425F24Rik; 6330416D14Rik; Als2cr13; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 13; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 13 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 13 protein homolog; FAM117B; family with sequence similarity 117 member B; family with sequence similarity 117, member B; protein FAM117B
Gene Symbols FAM117B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM117B. Recombinant protein control fragment (Product #RP-101060).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.