missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM117A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15732531
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15732531 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15732531 Supplier Invitrogen™ Supplier No. PA554598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KEISKLKQQL QRTKLSRSGK EKERGSPLLG DHAVRGALRA SPPSFPSGSP VLRLSPCLHR SLEGLNQELE EVFVKEQGEE ELLRILDIPD G Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

FAM117A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM117A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM117A
Gene Accession No. Q9C073
Gene Alias 5730593F17Rik; AW548096; C/EBP induced protein; C/EBP-induced protein; Fam117a; family with sequence similarity 117 member A; family with sequence similarity 117, member A; protein FAM117A; RGD1561062
Gene Symbols FAM117A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM117A. Recombinant protein control fragment (Product #RP-93239).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.