missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM115C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532110
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, K562, THP-1. Predicted reactivity: Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The FAM115C gene (Family with Sequence Similarity 115 Member C) is a relatively under-characterized gene located on chromosome 9q34.13. This gene encodes a protein that is part of the FAM115 family, which is involved in various cellular processes, including signal transduction, cellular growth, and maintenance. While detailed functional studies on FAM115C are limited, proteins in this family are often characterized by their involvement in cellular homeostasis and regulatory mechanisms. Research on FAM115C is nascent, but it is suggested to play a role in the central nervous system based on expression patterns in brain tissues.
Specifications
| FAM115C | |
| Polyclonal | |
| Unconjugated | |
| TCAF2 | |
| FAM115C; FAM139A; family with sequence similarity 115, member C; family with sequence similarity 139, member A; protein FAM115C; TCAF2; TRP channel-associated factor 2; TRPM8 channel associated factor 2; TRPM8 channel-associated factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285966 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.84 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| A6NFQ2 | |
| TCAF2 | |
| Recombinant protein encompassing a sequence within the center region of human FAM115C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction