missing translation for 'onlineSavingsMsg'
Learn More

FAM111B Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-18411R

Product Code. 18589879

  • £511.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FAM111B Polyclonal antibody specifically detects FAM111B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM111B
Polyclonal
Western Blot
family with sequence similarity 111, member B
Recognizes region between residues Between between 225 and 275..
0.1 mL
Primary
Human
Purified
Western Blot
DyLight 550
50mM Sodium Borate
Rabbit
Affinity purified
RUO
374393
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.