missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM111A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558827
This item is not returnable.
View return policy
Description
Immunogen sequence: RSRKHSVNEK CNMKIEHYFS PVSKEQQNNC STSLMRMESR GDPRATTNTQ AQRFHSPKKN PEDQTMPQNR TIYVTLKVNH RRNQDMKL Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 47%.
With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and isconsidered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene isimportant for regulation ofcellcycle arrest and apoptosisfollowing double strand DNA breaks. Atmmutation leadsto the disorderknown as ataxia telangiectasia. The blood disorders Sickle cell anemia and β thalassemia are caused by HBB gene mutations. Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitzsyndrome are also associatedwith defectsin chromosome 11. The FAM111A gene product has been provisionally designated FAM111A pending furthercharacterization.
Specifications
| FAM111A | |
| Polyclonal | |
| Unconjugated | |
| FAM111A | |
| 4632417K18Rik; AW413625; Fam111a; family with sequence similarity 111 member A; family with sequence similarity 111, member A; GCLEB; KCS2; Kiaa1895; Protein FAM111A; RGD1560913 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PZ2 | |
| FAM111A | |
| Recombinant protein corresponding to Human FAM111A. Recombinant protein control fragment (Product #RP-97448). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction