missing translation for 'onlineSavingsMsg'
Learn More

FAM111A Antibody, Novus Biologicals™

Product Code. 18401459 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401459 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401459 Supplier Novus Biologicals Supplier No. H00063901B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

FAM111A Polyclonal antibody specifically detects FAM111A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM111A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_071357.2
Gene Alias DKFZp686A06175, family with sequence similarity 111, member A, hypothetical protein LOC63901, KIAA1895FLJ22794
Host Species Mouse
Immunogen FAM111A (NP_071357.2, 1 a.a. - 611 a.a.) full-length human protein. MSCKKQRSRKHSVNEKCNMKIEHYFSPVSKEQQNNCSTSLMRMESRGDPRATTNTQAQRFHSPKKNPEDQTMPQNRTIYVTLKVNHRRNQDMKLKLTHSENSSLYMALNTLQAVRKEIETHQGQEMLVRGTEGIKEYINLGMPLSCFPEGGQVVITFSQSKSKQKEDNHIFGRQDKASTECVKFYIHAIGIGKCKRRIVKCGKLHKKGRKLCVYAFKGETIKDALCKDGRFLSFLENDDWKLIENNDTILESTQPVDELEGRYFQVEVEKRMVPSAAASQNPESEKRNTCVLREQIVAQYPSLKRESEKIIENFKKKMKVKNGETLFELHRTTFGKVTKNSSSIKVVKLLVRLSDSVGYLFWDSATTGYATCFVFKGLFILTCRHVIDSIVGDGIEPSKWATIIGQCVRVTFGYEELKDKETNYFFVEPWFEIHNEELDYAVLKLKENGQQVPMELYNGITPVPLSGLIHIIGHPYGEKKQIDACAVIPQGQRAKKCQERVQSKKAESPEYVHMYTQRSFQKIVHNPDVITYDTEFFFGASGSPVFDSKGSLVAMHAAGFAYTYQNETRSIIEFGSTMESILLDIKQRHKPWYEEVFVNQQDVEMMSDEDL
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63901
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.