missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM107A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35250-20ul
This item is not returnable.
View return policy
Description
FAM107A Polyclonal antibody specifically detects FAM107A in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| FAM107A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| downregulated in renal cell carcinoma, DRR1family with sequence similarity 107 member A transcript, family with sequence similarity 107, member A, FLJ30158, FLJ45473, Protein TU3A, TU3ADown-regulated in renal cell carcinoma 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human FAM107A (NP_009108.1).,, Sequence:, MYSEIQRERADIGGLMARPEYREWNPELIKPKKLLNPVKASRSHQELHRELLMNHRRGLGVDSKPELQRVLEHRRRNQLIKKKKEELEAKRLQCPFEQELLRRQQRLNQLEKPPEKEEDHAPEFIKVRENLRRIATLTSEEREL | |
| 20 μL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 11170 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction