missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM105A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598912
This item is not returnable.
View return policy
Description
FAM105A Polyclonal Antibody for Western Blot, IHC (P), ELISA
FAM105A belongs to the FAM105 family. The exact function of FAM105A remains unknown.
Specifications
| FAM105A | |
| Polyclonal | |
| Unconjugated | |
| FAM105A | |
| 9830126M18; FAM105A; family with sequence similarity 105 member A; family with sequence similarity 105, member A; inactive ubiquitin thioesterase FAM105A; Inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase FAM105A; LOW QUALITY PROTEIN: inactive ubiquitin thioesterase FAM105A; NET20; OTU deubiquitinase with linear linkage specificity like; OTULINL; protein FAM105A; Protein FAM105A-like protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54491 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9NUU6 | |
| FAM105A | |
| Recombinant Human Inactive ubiquitin thioesterase FAM105A protein (81-356AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction